Balances, triple beam with Dial-O-Gram®, 1600 series
Lieferant: OHAUS
80000006
611-5004EA 493 EUR
611-5004
Balances, triple beam with Dial-O-Gram®, 1600 series
Waagen
Triple beam balance that provides high quality, accuracy and complete flexibility in the classroom.
  • Metal base, beam and pan made of stainless steel
  • Magnetic damping provides three times faster stability reading
Features counterbalancing knob for quick zeroing, positive poise positioning for repeatable results, self-aligning beam design, magnetic damping to minimise oscillation and speed weighing.