Cubis® II Balances
Lieferant: Sartorius Balances
MCA125S-U-MTS
611-5824EA 14010 EUR
611-5824
Cubis® II Balances
Waagen
The modular weighing system Cubis® II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks.
  • Leading performance
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-levelling
  • Full end-to-end data integrity
More Product Information
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Operating temperature range 0...40 °C
Environmentally Preferable
Plug type Two-sided plug
Kommunikationsschnittstelle COM-RS232 interface
Spannung 0 - 240 VAC (±10%)