Dynamometers / spring balances, 281 series
Lieferant: KERN SOHN
281-601
611-8068EA 105 EUR
611-8068
Dynamometers / spring balances, 281 series
Waagen
Precise, mechanical spring balances in robust aluminium housing with g/kg readability.
  • Aluminium scale tube: Robust, long service life, rustproof
  • Compressive force measurement: Possible using an optional pressure set
  • Thanks to the rotating handrail the scale can always be aligned to be at the very best line of sight
  • Non-fatigue stainless steel spring
  • Clip loop can be freely rotated of the lower suspension bracket by 360° for models with max. capacity ≤ 1 kg
Wear-resistant, colour-anodised precision scale with high resolution for accurate readout of the measuring result.