Plane weighing glass, according to Heidbrink
Lieferant: WITEG LABORTECHNIK
2.873.902
611-9090EA 81.6 EUR
611-9090
Plane weighing glass, according to Heidbrink
Schiffchen Wägeschiffchen
Borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
For all weighings in which there is an advantage when the sample is divided in thin layers.