Weighing boats, aluminium
Lieferant: HEATHROW
HS14521A
611-9111EA 14.7 EUR
611-9111
Weighing boats, aluminium
Schiffchen Wägeschiffchen
These aluminium weighing boats are sturdy yet lightweight.
  • Boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity
  • Finger tab handle can be etched for identification prior to weighing
  • Disposable
They can be used for general-purpose weighing, dispensing, storage, food processing, evaporating, drying, or a great dust cover for open vessels.
Standards: FDA/food-grade compliant.
Verpackung: Packs of 100.