Insulin syringes, Omnican® 20
Lieferant: B. Braun
9161619S
612-0140EA 85.6 EUR
612-0140
Insulin syringes, Omnican® 20
Spritzen Spritzen, medizinisch
Insulin syringe made of PP/PS with round shaped cylinder with black scale.
  • Piston with double sealing ring
  • Protective caps at the ends of the syringe for sterility
  • Integral piston stops
  • Latex-free
The crystal-clear barrel of the syringe makes sure that air bubbles are easily identified.