Burettes, straight stopcock, Schellbach, FORTUNA®
Lieferant: Poulten & Graf
1.0083402G
612-0564EA 53.4 EUR
612-0564
Burettes, straight stopcock, Schellbach, FORTUNA®
Büretten
Borosilicate glass 3.3, class AS.
  • Calibrated to deliver 'Ex'
  • Schellbach stripe, blue enamel graduations
*Short form
Standards: EN ISO 385