Burettes, straight PTFE stopcock, class A, Volac®
Lieferant: Poulten & Graf
RB9806/NC/I
612-1698EA 95.8 EUR
612-1698
Burettes, straight PTFE stopcock, class A, Volac®
Büretten
Burette, Class A, made of borosilicate glass, with stopcock with interchangeable PTFE key.
  • Clearly legible lettering and fine graduations lines in white color
  • Wide hopper for easy filling
  • Interchangeable glass spikes for better association
Class A burettes are individually calibrated, tested at 3 points.