Rack for pipetting pumps
Lieferant: DELTALAB
W-150
612-2080EA 0 EUR
612-2080
Rack for pipetting pumps
Pipettenständer
Pipette pump support rack made of acrylic resin and designed to keep in position four pipette pumps.
  • Angled for quick placement and removal of pipettors
  • U-shaped channels and a lower deck provide secure and convenient storage
  • Suitable for pipette pumps W-100, W-110, W-120