Pipettierhelfer, pipetus®-junior
Lieferant: HIRSCHMANN
9902000
612-2604EA 160 EUR
612-2604
Pipettierhelfer, pipetus®-junior
Pipetten Pipettierhelfer
Pipette controller for glass and plastic pipettes from 0,5 to 25 ml.
  • Easy, ergonomic, safe and accurate
  • Total expulsion of the liquid simply by pressing the button on the back of the instrument
  • The integrated, hydrophobic PTFE membrane filter ensures the pipette is protected against accidental overfilling
Full pressure on the fill button allows suction of around 2 ml. By pressing on the empty button, the pipette is emptied gravimetrically and the degree of pressure on the button determines the rate of flow.