Measuring cylinders, tall form, class B, borosilicate glass, SILBERBRAND ETERNA
Lieferant: Brand
31962 31964 31920 31905 31908 31928 31938 31948 31954
612-5014EA 61.9 EUR
612-5014 612-4218 612-5003 612-5000 612-5002 612-5004 612-5011 612-5012 612-5013
Measuring cylinders, tall form, class B, borosilicate glass, SILBERBRAND ETERNA
Zylinder Messzylinder
Borosilikatglas 3.3, hohe Form, Klasse B.
  • Calibrated to contain (TC, In)
  • Hexagonal base, spout
  • Short graduation marks
  • Marks and inscriptions in ETERNA amber stain
DIN EN ISO 4788
Standards: DIN EN ISO 4788.