VWR®, Measuring Cylinders, Tall Form, Class B, PMP TPX
Lieferant: VWR Collection
612-5572EA 2.9 EUR
612-5572 612-5573 612-5579 612-5587 612-5589 612-5596 612-5599 612-5601
VWR®, Measuring Cylinders, Tall Form, Class B, PMP TPX
Zylinder Messzylinder
PMP (TPX), crystal-clear, tall form, class B.
  • Stable hexagonal base
  • Graduations are calibrated in mould for accuracy and consistency
  • Excellent chemical resistance
  • Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C short period)
  • Resin meets US FDA 21 CFR requirements
Standards: DIN 12681/ISO 6706