WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
Lieferant: DWK Life Sciences
W155091
612-6770EA 924 EUR
612-6770
WHEATON® PIPET-PAL® Pipette Controller, Translucent Blue
Pipetten Pipettierhelfer
PIPET-PAL® pipette controller are lightweight (195 g) and well-balanced, making even prolonged pipetting very comfortable.
  • Quiet, robust pump for high pipetting speed
  • Versatile for both fine dosing and gravimetric dispensing
  • Eliminate contamination with a replaceable, sterile filter
  • Lithium-Polymer battery technology for long operating time
  • Autoclavable pipette holder for cleanliness and safety
  • One year warranty
Precise finger-control of pipetting enables accurate dispensing, even drop-by drop.
Designed for use with all serological pipettes in the volume range 1 to 100 ml.