Einkanalpipetten, elektronisch, variables Volumen, Eppendorf Xplorer®
Lieferant: EPPENDORF
4861000.015 4861000.040 4861000.023 4861000.031 4861000.058 4861000.066 4861000.027 4861000.044 4861000.017
613-1398EA 714 EUR
613-1398 613-1407 613-1399 613-1400 613-1408 613-1409 613-6981 613-6982 613-6980
Einkanalpipetten, elektronisch, variables Volumen, Eppendorf Xplorer®
Pipetten Mikropipetten
Eppendorf Xplorer single-channel pipettes provide reliable, ergonomic high-precision liquid handling. With easy operation, five pipetting modes, and advanced charging technology, they minimise errors and boost reproducibility in a range of lab applications.
  • Electronic single channel pipettes designed to aspirate and dispense liquid volumes from 0,5 μl to 10 ml
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker with ergonomic display angle
  • Selection dial and innovative ejector
  • Convenient handle and powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 and 10 ml)
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish. Air-cushion pipetting system, with autoclavable lower part and epT.I.P.S.® consumables.
Applications: Pipetting of aqueous liquids.
Bestellinformation: Supplied with charger.