Pipette controller, Pipet-Aid®
Lieferant: DRUMMOND
4-000-221-TC 4-000-221
613-4255EA 1200 EUR
613-4255 DRUM4-000-221
Pipette controller, Pipet-Aid®
Pipetten Pipettierhelfer
This controller with easy-to-use volume control mechanism permits precise fluid intake and delivery. Can be used with glass or plastic pipettes.
  • Check valve protects against accidental fluid intake into the handle and
  • Special filtration unit for use with biohazardous materials is available
  • Dual pump filtration unit has aretention efficiency of 99,9999+% for 0,1 µm particles and droplets
  • Optional Tissue Culture (TC) nosepiece will prevent cross-contamination due to over-pipetting