Pipette stand, rotary
Lieferant: Sartorius
LH-725630
613-5492EA 139 EUR
613-5492
Pipette stand, rotary
Pipettenständer
Carousel stand for six mechanical pipettes.
When the pipette is not in use, it should be stored in an upright position in order to avoid contamination from desks and benches. This compact carousel stand is ideal for saving bench space and is compatible with all mechanical pipettes.