Filter tips, Corning® IsoTip™
Lieferant: Corning
4807 4808 4809 4810 4821 4823 4801
613-5506EA 149.11 EUR
613-5506 613-5507 613-5508 613-5509 613-5511 613-5512 613-5505
Filter tips, Corning® IsoTip™
Pipettenspitzen
These pipette tips feature an inert, hydrophobic bonded-fiber barrier that prevents aerosolised contaminants from coming in contact with air displacement pipettors. Ideal for applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Exceptional durability
  • All Corning® tip racks stand up to the demands of use with multichannel pipettors and withstand repeated autoclaving cycles
  • Sterile and certified RNase-/ DNase-free and non pyrogenic