3,3',5,5'-Tetramethylbenzidin (TMB), unlöslich, Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore (Oncogene)

Synonyme: TMB

613548-100
613548-100EA 167 EUR
613548-100
3,3',5,5'-Tetramethylbenzidin (TMB), unlöslich, Calbiochem®, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin (TMB)

Peroxidase substrate that produces a stable, insoluble, aquamarine blue precipitate at reaction site with little or no background.


Formulation: 1.19 mM H₂O₂ in a proprietary solvent, 1.13 mM TMB, pH 4.9 ± 0.03.

Formel: C₁₆H₂₀N₂
Siedepunkt: 402 °C (1013 hPa)
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007748
CAS-Nummer: 54827-17-7
EINECS: 259-364-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR