Precision Alcohol Meters
Lieferant: Alla France
0910FC000/20QP 0910FC010/20QP 0910FC020/20QP 0910FC030/20QP 0910FC040/20QP 0910FC050/20QP 091.060FCP 0910FC080/20QP 0910FC090/20QP 0910FC005/20QP 0910FC006/20QP 0910FC045/20QP 0910FC055/20QP 0910FC065/20QP 0910FC075/20QP 0910FC095/20QP 0920FC000/20QP 0920FC020/20QP 0920FC040/20QP 0920FC060/20QP 0920FC080/20QP
614-0004EA 178 EUR
614-0004 614-0006 614-0007 ALLA0910FC030/20QP ALLA0910FC040/20QP 614-0013 614-0015 614-0021 614-0022 ALLA0910FC005/20QP ALLA0910FC006/20QP ALLA0910FC045/20QP ALLA0910FC055/20QP ALLA0910FC065/20QP ALLA0910FC075/20QP ALLA0910FC095/20QP ALLA0920FC000/20QP ALLA0920FC020/20QP ALLA0920FC040/20QP ALLA0920FC060/20QP ALLA0920FC080/20QP
Precision Alcohol Meters
Aräometer
These high precision alcoholometers with individual number on the scale and a calibration temperature of 20 °C.
Standards: ISO 4801 and NF B 35-515
Lieferumfang: Supplied with a certificate of conformity.