Standard-Aräometer
Lieferant: Alla France
3500LC07020BQP 3500LC07520BQP 3500LC08020BQP 3500LC08520BQP 3500LC09020BQP 3500LC09520BQP 3500LC10020MQP 3500LC10520MQP 3500LC11020MQP 350L115ISO649M 3500LC12020MQP 3500LC12520MQP 3500LC13020HQP 3500LC13520HQP
ALLA3500LC07020BQPEA 58.3 EUR
ALLA3500LC07020BQP ALLA3500LC07520BQP 614-1537 614-1539 614-1541 614-1543 614-1545 614-1547 614-1549 614-1551 614-1553 614-1554 614-1555 614-1556
Standard-Aräometer
Aräometer
These standard hydrometers strictly adhere to the principles of manufacture, and specified calibration.
  • Reading below meniscus
  • Calibration temperature: 20 °C
  • Reference mark on the stem shows any movement of the measurement scale
Standards: NF, ISO, BS and DIN
Bestellinformation: Include three calibration points (factory certificate or accredited laboratory certificate on request. The calibration points should be specified when ordering.
Lieferumfang: Supplied with certificate of conformity to ISO 649 (corresponding to BS718, DIN12791, NF B 35511).