TAK-242 ≥98% (durch HPLC), Calbiochem®

Lieferant: Merck Millipore Calbiochem
Achtung

Synonyme: Resatorvid, (R)-Ethyl 6-(N-(2-chloro-4-fluorophenyl)sulphamoyl)cyclohex-1-enecarboxylate, 6R-[[(2-chloro-4-fluorophenyl)amino]sulphonyl]-1-cyclohexene-1-carboxylic acid, ethyl ester

Verpackt unter Schutzgas

614316-5MG
614316-5MGEA 230 EUR
614316-5MG
TAK-242 ≥98% (durch HPLC), Calbiochem®
TAK-242

TAK-242, TLR4 inhibitor is a cell-permeable compound that selectively binds to Cys747 of TLR4 and selectively disrupts its interaction with adaptor molecules TIRAP and TRAM.

Formel: C₁₅H₁₇NO₄ClFS
Molecular Weight: 361,8 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11046362
CAS-Nummer: 243984-11-4

Bestellen

Spezifikation Testergebnisse

Purity ≥98% by HPLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR