Falling ball viscometer
Lieferant: HAAKE
356-0001
616-3282EA 3050 EUR
616-3282
Falling ball viscometer
Viskosimeter Kugelfall-Viskosimeter
This falling ball viscometer provides an accurate way of measuring the viscosity, in transparent Newtonian liquids and gases.
Specially suited for the chemical (polymer solutions, solvents, inks), pharmaceutical (raw materials, glycerine), food (gelatin, sugar solutions) and mineral oil (oils, liquid hydrocarbons) industries.
Standards: DIN 53015, ISO 12058
Lieferumfang: Supplied with six balls, instrument case, thermometer, cleaning tools, calibration sheet and instruction manual.