Calbiochem® Human Transferrin (apo) (from plasma)
Lieferant: Merck Millipore Calbiochem
616395-100
616395-100EA 163 EUR
616395-100
Calbiochem® Human Transferrin (apo) (from plasma)
Proteine und Peptide
Native apo-transferrin from human plasma.
  • Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
Transferrin is a serum protein involved in iron binding and transport. Substantially iron-free.