Alkohol 94-96, AR® ACS (IPA 5% + MeoH 5%), Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Reagent alcohol

6183-10EA 248.05 EUR
6183-10
Alkohol 94-96, AR® ACS (IPA 5% + MeoH 5%), Macron Fine Chemicals™
Alcohol

Made from specially denatured alcohol formula 3A. SDA formula 3A consists of 5 volumes of methanol and 100 volumes of 200 proof ethanol.


  • Total alcohol: 99.5%
  • Ethyl alcohol: 89 to 92%
  • Denaturant: 3.5 to 5.5% methyl alcohol and 4 to 6% 2-propanol


Bestellinformation: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 94.0 - 96.0%
For Laboratory,Research,or Manufacturing Use 3.5 - 5.5%
Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Methanol (v/v) 3.5 - 5.5 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Specific Gravity at 25°C 0.782 - 0.793
Water (H₂O) ≤0.5 %
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR