Thermometer clamp, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F182410000
620-1030EA 48.8 EUR
620-1030
Thermometer clamp, SP Bel-Art
Klemmen Klemmen für den allgemeinen Gebrauch
These PP clips hold thermometers and probes from 3,2 to 11 mm in diameter securely in place.
  • Fits onto the rims of beakers or pitchers up to ¼” thick
  • Autoclavable at 121 °C