Kernthermometer, TTX 110
Lieferant: EBRO
1340-5110
620-1384EA 70 EUR
620-1384
Kernthermometer, TTX 110
Thermometer Digitalthermometer
Thermocouple thermometer with fixed probe (Type T).
  • Fast measurements, high accuracy
  • HACCP conform
  • Robust and impact resistant
  • Replaceable battery
  • Automatic switch-off
  • HOLD function
Ideal for use in the food industry, transport and storage applications.
EN 13485, conform to HACCP
Standards: Conforms to HACCP and EN 13485.
Lieferumfang: Supplied with a replaceable battery and factory calibration certificate.