Digital thermo-hygrometer, TFH 620
Lieferant: EBRO
1340-5621
620-1394EA 411 EUR
620-1394
Digital thermo-hygrometer, TFH 620
Hygrometer Thermohygrometer, digital
This robust and impact resistant thermo-hygrometer provides high accuracy measurements for monitoring storage checks.
  • Automatic deactivation
  • Capacitive humidity sensor
  • Two measurement channels
Thermistor sensor.
Lieferumfang: Supplied with an internal sensor, a six-point factory calibration certificate, a lithium three-volt battery for up to five years of life with a charge indicator.