Core thermometer, TDC 150
Lieferant: EBRO
1340-1611
620-1568EA 45.9 EUR
620-1568
Core thermometer, TDC 150
Thermometer Digitalthermometer
Core thermometer works with fast response time.
  • Fixed probe, pointed
  • Robust and impact-resistant
  • Replaceable battery
  • Switchable between °C/°F