Thermometer-Halter, Labo-Clip
Lieferant: AMARELL, ARNO
K160310
620-1999EA 49.44 EUR
620-1999
Thermometer-Halter, Labo-Clip
Thermometer-Halterungen
This spring clip holder can be attached quickly for holding thermometers, test tubes and pH meter electrodes. Stainless steel.
  • Rubber base
  • Strudy design
  • Circular support
  • Can be fixed on any container
  • Five holes of different Ø: 4; 5; 6; 10 and 13 mm
Maximum length: 12 mm.