VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 127 mm Probe
Lieferant: VWR Collection
620-2133EA 55.8 EUR
620-2133
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 127 mm Probe
Thermometer Digitalthermometer
Digital stem thermometer suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Standards: Tested and calibrated in exclusive triple-accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448,01), against equipment whose calibration is traceable to NIST.
Lieferumfang: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish).
Bitte beachten These electronic stem thermometers are not suitable for solids such as frozen items.