VWR® Traceable®, Digitale Thermo-Hygrometer mit externen Fühlern
Lieferant: VWR Collection
620-1874EA 272 EUR
620-1874 620-2273
VWR® Traceable®, Digitale Thermo-Hygrometer mit externen Fühlern
Hygrometer Thermohygrometer, digital
This thermo-hygrometer measures temperature and relative humidity displaying both on a dual-LCD. Designed for monitoring conditions in cleanrooms, computer rooms, storage facilities and anywhere temperature and RH affect vital components and tests.
  • Maintains accuracy with 'do it yourself' calibration
  • Simultaneously displays both temperature and relative humidity on dual-display LCD (28,6 mm character height)
  • Minimum/maximum memory
  • Interface for transfer of data to a computer
  • An audible click confirms each key entry
Compliant ISO/IEC 17025, ANSI/NSCL Z5More Product Information