Pocket thermometer, QUICKFIT®
Lieferant: SciLabware
SH4A
620-2286EA 28.8 EUR
620-2286
Pocket thermometer, QUICKFIT®
Thermometer Analoge Thermometer
Quickfit® ground-glass joints are precision manufactured with an accurate taper and smooth finish for leak-free performance and assured interchangeability.
  • For incorporating thermometers into Quickfit® jointed glassware assemblies
  • Pocket fits most Quickfit® still heads