VWR® Digital Barometer Data Acquisition System
Lieferant: VWR Collection
620-2331EA 207 EUR
620-2331
VWR® Digital Barometer Data Acquisition System
Software Labor-Software
Ideal for monitoring environmental conditions in plants and labs.
  • Set high and low alarm points
  • Connect and download real-time readings to a PC
  • Get reliable readings
Use this data acquisition software and USB cable to permanently capture real-time measurements on a PC from up to eight different Traceable® barometers. Data can then be saved, viewed, printed as-is, or merged into a report or spreadsheet and emailed to any smartphone or computer. The software also features a 24/7 audible and visual alarm to remind users to check on data.
Traceable® products have an individually numbered Traceable® Certificate provided with each unit, that assures accuracy from an ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable® name, thus saving users both time and money by not having to calibrate separately.
Lieferumfang: Includes NIST-traceable certificate from A2LA accredited (1750.01) calibration laboratory.