VWR® Traceable®, Infrared Thermometer, with Wristband
Lieferant: VWR Collection
620-2663EA 119 EUR
620-2663
VWR® Traceable®, Infrared Thermometer, with Wristband
Thermometer Digitalthermometer
Traceable® infrared laser thermometer is simple to operate - turn on, point at sample, and take a reading in less than a second. Unit reads both Fahrenheit and Celsius of any surface - solids, semi-solids, and liquids. Non invasive, no touch measurements.
  • Field of view is 6:1
  • Emissivity is 0,05 to 1,00 in 0,01 steps
  • Minimum and maximum feature allows the viewing of highest and lowest readings at any time
  • Laser beam allows perfect sample target sighting
  • Handy wristband
  • Display indicates low battery
Ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics and field use.
Standards: To ensure accuracy, an individually serial numbered Traceable® certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Lieferumfang: Supplied with battery, wristband and Traceable® certificate.