VWR® Traceable®, Indoor/Outdoor Digital Thermometer with Giant Dual-Display and Calibration
Lieferant: VWR Collection
620-2713EA 75.6 EUR
620-2713
VWR® Traceable®, Indoor/Outdoor Digital Thermometer with Giant Dual-Display and Calibration
Thermometer Digitalthermometer
This thermometer features a jumbo digital display which is easy to read from distances.
  • Dual display permits reading ambient and probe temperatures simultaneously
  • °F/°C switchable
  • HI/LO Alarms and MIN/MAX memory readings may be displayed and reset at the touch of a key
  • Cable and sensor perform accurately when underwater
  • Always-on instrument reads temperature constantly
Standards: NIST-traceable calibration also helps meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.