Temperature probe, TPC 300H, for EBI 300 TE
Lieferant: EBRO
1341-6370
620-2862EA 62 EUR
620-2862
Temperature probe, TPC 300H, for EBI 300 TE
Temperaturfühler
Calibrated replacement probe with handle for EBI 300 TE data logger.
  • NTC sensor
  • IP65 protection class
  • Fast, flexible core temperature measurements
Measuring range: −35...+75 °C