pH electrode for general applications, gel-glass
Lieferant: HAMILTON BONADUZ
238025
620-4908EA 152 EUR
620-4908
pH electrode for general applications, gel-glass
Elektroden pH-Elektroden
Gel-Glass is a maintenance-free combination electrode for everyday applications.
  • Ag/AgCl reference system
  • Cylindrical membrane