Lithium ionophore VI ≥99,0% (durch HPLC), Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyme: 6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane, 6,6-Dibenzyl-14-crown-4

M-Clarity™ Qualitätsstufe = MQ100

62567-100MG
62567-100MGEA 695 EUR
62567-100MG
Lithium ionophore VI ≥99,0% (durch HPLC), Selectophore™, Funktion getestet Ionophor, Supelco®
Lithium ionophore VI

Neutral carrier for Li+-ions in PVC ion-selective electrodes with excellent selectivities over potassium and sodium ions.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₄H₃₂O₄
Molecular Weight: 384,51 g/mol
MDL: MFCD00037498
CAS-Nummer: 106868-21-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad703b18ab3ef107770fd1e4a0943166fbdf3252&vid=06106bf5e26d845012024132d6f8520519000f91&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR