Immersion oils
Lieferant: ZEISS
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
- Bubble-free immersion oil
- Suitable for oil immersion objectives from Carl Zeiss
Standards: ISO 8036-1/2