Zählkammern, Bürker
Lieferant: MARIENFELD
0640211 0640210 0640231 0640331 0640310 0640311 0640330 0640230
630-1541EA 56.15 EUR
630-1541 631-0920 630-1542 630-1545 630-1543 631-0922 630-1544 MARI0640230
Zählkammern, Bürker
Objektträger Zählkammern
Glass counting chambers equipped with two sets of grids, engraved into two ground, and polished surfaces on the central ridge. This central ridge is located between two elevated ridges, that are also ground and polished.
Designed for investigations of cell suspensions.
Conform to DIN 12847 and the German Calibration Ordinance
Standards: Konform mit DIN 12847 und der Deutschen Kalibrierverordnung
Lieferumfang: Wird in durchsichtiger Kunststoffbox geliefert.