Handheld digital microscopes, Dino-Lite Long Working Distance (LWD)
Lieferant: Dino-Lite / IDCP B.V.
AM4013MZTL AM4013MTL AM4113TL AM4113TL-M40 AM4113ZTL AM4115TL AM4115ZTL AM4815ZTL AM4515ZTL AM7515MZTL AM7115MZTL AM7915MZTL AM4115TF
630-1346EA 619 EUR
630-1346 630-2651 630-1617 630-2652 630-2653 630-2514 630-2515 630-2655 630-2654 630-2659 630-2657 630-2753 IDCPAM4115TF
Handheld digital microscopes, Dino-Lite Long Working Distance (LWD)
Mikroskope
These microscopes are ideal for tasks where extra distance to the object and a larger field of view is required. Magnification range: 5 to 140×.
  • High speed USB 2.0 connection
  • Long working distance of up to 22,5 cm (TF models up to 43 cm)
  • Metal housing or composite housing
  • Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser
More Product Information