Digital magnifier, Mobilux®
Lieferant: ESCHENBACH OPTIK
1651-101
630-2716EA 0 EUR
630-2716
Digital magnifier, Mobilux®
Lupen
The Mobilux® is a portable video magnifier featuring a 4,3" HD TFT LCD screen with anti-glare coating and LED illumination for digital inspection.
  • Smaller steps and continuous zoom are also possible
  • Three brightness settings, adjustable lines and blinds for precise page orientation
  • Various tone settings ensure a comfortable visual experience
It provides time and date and has an automatic shut-off feature after five minutes of non-use and five customisation modes.
A 4 GB SD card stores photos, which are downloadable to a PC or Mac computer through the included USB connection (Windows 10 compatible).
Mobilux® Digital Touch HD software version 2.0 upgrade is available.
Lieferumfang: Supplied in a zippered crush-proof protective case is included, along with a cleaning cloth and wrist strap.