Cover glasses
Lieferant: GLASWARENFABRIK KARL HECHT
41014542
630-2746EA 193 EUR
630-2746
Cover glasses
Deckgläser
Cover glasses with controlled thickness.
  • Free from colour distortion borosilicate glass D 263™ M
  • Excellent quality: Low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
Standards: ISO 8255
Verpackung: In tight-closing plastic boxes for protection against damp and climatic influences.