Square cover glasses for fluorescence
Lieferant: MARIENFELD
0102032 0100032 0100052 0100062 0102052 0102062 0103032 0103052 0103062 0102042 0100042
630-2758EA 41.4 EUR
630-2758 630-2754 630-2756 630-2757 630-2760 630-2761 630-2762 630-2763 630-2764 630-2759 630-2755
Square cover glasses for fluorescence
Deckgläser
These cover glasses are made of borosilicate glass D 263™ M of the hydrolytic class 1.
  • Colourless and clear
  • Chemically resistant
Suitable for fluorescence microscopy.
Standards: ISO 8255 - Approved for in vitro diagnostic (IVD) applications according to IVD directive 98/79/EC.
Verpackung: In hinged lid plastic boxes of 100 or 200 pieces - Ten boxes in a carton.