Magnifier, Linen Tester
Lieferant: ESCHENBACH OPTIK
12003
630-8321EA 16.5 EUR
630-8321
Magnifier, Linen Tester
Lupen
Thread counter with side scale, measuring thread counter made of plastic. Available for the metal and plastic thread counters.
  • Division of the scale in millimeters
  • Plano-convex PXM® lightweight lens, DUPLEX coated for sharp-edged, distortion-free images
  • Black plastic housing