Dow Corning® FS 1265/10.000 cS chemisch inerte Flüssigkeit
Lieferant: DOW CORNING
630267H
630267HEA
0
EUR
630267H
Dow Corning® FS 1265/10.000 cS chemisch inerte Flüssigkeit
Polyfluorsiloxan, kinematische Viskosität 10.000 cSt (25°C)
Clear low volatility fluorosilicone especially suitable for foam control in chlorinated solvents
Lieferumfang: This product is not available in all countries. Please check with your local VWR International office or supplier.
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
63148-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...