Magnesiumperchlorat Hexahydrat, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

63043-50ML-F
63043-50ML-FEA 100 EUR
63043-50ML-F
Magnesiumperchlorat Hexahydrat, Supelco®
Magnesiumperchlorat Hexahydrat
Formel: Cl₂MgO₈·6H₂O
Molecular Weight: 331,3 g/mol
Schmelzpunkt: 185…190 °C
Dichte: 1,98 g/cm³ (20 °C)
MDL: MFCD00149778
CAS-Nummer: 13446-19-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR