Magnesium ionophore III, Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
Ionophore used in solvent polymeric membrane electrodes for the assay of Mg2+ activities in blood serum
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₃₀H₅₈N₄O₄ |
MDL:
MFCD00144715 CAS-Nummer: 119110-38-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...