Magnesium ionophore IV, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

63088-1EA
63088-1EAEA 1290 EUR
63088-1EA
Magnesium ionophore IV, Selectophore™, Funktion getestet Ionophor, Supelco®
Magnesium ionophore IV

Very lipophilic magnesium ionophore; assay of Mg2+-activity in human blood serum with an optimized liquid polymer membrane electrode


50 mg solution in 0.5 mL tetrahydrofuran


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₄₉H₉₄N₆O₆
MDL: MFCD00144716
CAS-Nummer: 135734-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c922ae1dab31ce0dece36fba1b1820f68dd427d0&vid=bbd911d39345855c14dbe0fdaff516f6d294aaa0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR