Magnesiumoxid light 99, tested according to Ph.Eur., Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
gebrannte Magnesia
63096-500G
63096-500GEA
202.39
EUR
63096-500G
Magnesiumoxid light 99, tested according to Ph.Eur., Fluka™
Magnesiumoxid
Formel:
MgO Molecular Weight: 40.3 g/mol Siedepunkt: 3.600 °C (1.013 hPa) Schmelzpunkt: 2.852 °C Dichte: 3.58 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011109 CAS-Nummer: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Spezifikation Testergebnisse
Identity B | complying |
Assay (calculated to ignited substance) | 98.0 - 100.5 % |
Appearance of the solution | complying |
Loss on ignition | Max. 8.0 % |
In water soluble matter | Max. 2.0 % |
Insoluble in acetic acid | Max. 0.1 % |
Arsenic (As) | Max. 4 ppm |
Calcium (Ca) | Max. 1.5 % |
Iron (Fe) | Max. 0.1 % |
Heavy metals (as Pb) | Max. 30 ppm |
Chloride (Cl) | Max. 0.15 % |
Sulfate (SO₄) | Max. 1.0 % |
Bulk Density | Max. 0.15 g/ml |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...